Learn More
GE Healthcare GeneQuant™ 1300 Spectrophotometers
Can be used with a wide variety of cuvettes or capillaries for sample volumes from 3µL to 2mL
Manufacturer:  GE Healthcare 28918215
 View more versions of this product
Catalog No. 45-001-536
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
GeneQuant 1300 Spectrophotometer with Bluetooth
CCD array
DNA, RNA and oligonucleotide purity and concentration measurements, quantitation of proteins, nucleic acids and bacterial cell cultures, cell density and enzyme kinetics, cDNA application software for measuring incorporation of CyTM3 and Cy5 or other dyes into purified microarray hybridization probes and PCR products
39cm
Xenon
100 to 240V
190 to 1100nm
9 lb.
3.93 in.
3 Year
100 to 240V 50/60Hz
15.35 in.
50/60Hz
5nm
±2nm
±0.5nm
4.5kg
10cm