Learn More
GE Healthcare Glass Plate Kit
Used when spot picking lab-cast gels and when background fluorescence is not concerned
Manufacturer:  GE Healthcare 80647539
Catalog No. 45-005-914
Add to cart
Includes:
  • 1 Glass plate (27.6 x 21.7cm x 3mm)
  • 1 Glass plate (27.6 x 21.1cm x 3mm)
  • 2 Vinyl spacer (10mm x 1mm)
Specifications
Specifications
1 Glass plate (27.6 x 21.7cm x 3mm), 1 Glass plate (27.6 x 21.1cm x 3mm), 2 Vinyl spacer (10 x 1mm)
Complete packsize
Glass Plate Kit