Learn More
GE Healthcare GST Detection Module
For identification of GST fusion proteins using either a biochemical or immunological assay; the GST Detection Module also includes a goat/anti GST polyclonal antibody suitable for use in Western blots
Manufacturer:  GE Healthcare 27459001
Catalog No. 45-001-378
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Uses either a biochemical or immunological assay
  • In the biochemical assay, glutathione and 1-chloro-2-4-dinitrobenzene (CDNB) serve as substrates for GST to yield a yellow product detectable at 340 nm
  • The immunoassay makes use of an affinity purified anti-GST antibody
Specifications
Specifications
Identifies GST fusion Proteins
GST detection module
50 Detections
2° to 8°C