Learn More
GE Healthcare GST Gene Fusion System pGEX Vectors
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schistosoma japonicum GST
Manufacturer:  GE Healthcare 28954646
 View more versions of this product
Catalog No. 45-500-127
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Offers a tac promoter for chemically inducible, high-level expression of GST-tagged recombinant proteins
  • Offers an internal laq1q gene for use in any E. coli host
  • With PreScission™ Protease, Thrombin, or Factor Xa recognition sites for cleaving the target protein from the fusion product
  • The vectors provide all three translational reading frames beginning with the EcoRI restriction site
  • Can use a wide variety of E. coli host strains cloning and expression
  • pGEX-1lT EcoRI/BAP is an EcoR1 pre cut vector, phosphatase treated to prevent self ligation, ready for gene insertion
  • pGEX-2TK will introduce a specific kinase site allowing the fusion protein to be labelled with [?-32P]ATP using protein kinase
Specifications
Specifications
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schisto soma japonicum GST
2TK
pGEX vecto r
25μg