GE Healthcare GST Gene Fusion System pGEX Vectors
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schistosoma japonicum GST
Manufacturer: GE Healthcare 28954654
 View more versions of this product
Catalog No. 45-500-128
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Includes 3X
    Product Type pGEX vecto r
    For Use With (Application) Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schisto soma japonicum GST
    Quantity 25μg
    • Offers a tac promoter for chemically inducible, high-level expression of GST-tagged recombinant proteins
    • Offers an internal laq1q gene for use in any E. coli host
    • With PreScission™ Protease, Thrombin, or Factor Xa recognition sites for cleaving the target protein from the fusion product
    • The vectors provide all three translational reading frames beginning with the EcoRI restriction site
    • Can use a wide variety of E. coli host strains cloning and expression
    • pGEX-1lT EcoRI/BAP is an EcoR1 pre cut vector, phosphatase treated to prevent self ligation, ready for gene insertion
    • pGEX-2TK will introduce a specific kinase site allowing the fusion protein to be labelled with [?-32P]ATP using protein kinase
  • Description & Specifications
Related Products
Product Content Feedback