Learn More
GE Healthcare GST Vector Primers for Sequencing Available on GSA/VA Contract for Federal Government customers only.
Forward and reverse sequencing primers flanking the multiple cloning site of all pGEX vectors in the GST fusion system, for verification of inserted DNA sequence
Manufacturer:  GE Healthcare 27141001
 View more versions of this product
Catalog No. 45-500-118
Add to cart
Description
Description
  • Provided ready for immediate use
  • pGEX 5’ Sequencing Primer: 5’-d[GGGCTGGCAAGCCACGTTTGGTG]-3’
  • pGEX 3' Sequencing Primer: 5’-d[CCGGGAGCTGCATGTGTCAGAGG]-3’
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Glutathione S-transferase (GST) gene fusion system
pGEX 5′ sequencing primer
-20°C
Ready-to-use in sequencing double-stranded DNA inserted into pGEX Vectors
260U