Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva GSTrap™ 4B Prepacked Columns: 16 x 25mm
For fast and simple purification of GST-tagged proteins
Supplier: Cytiva 28401747
Description
- Prepacked with Glutathione Sepharose™ 4B media, featuring a glutathione ligand coupled to 4% agarose
- May be used with syringe, peristaltic pump or ¨A¨KTA design chromatography system
- Purifies GST-tagged proteins from bacterial lysates and other glutathione S-transferases or glutathione-dependent proteins
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
- Average particle size: 90μm
- Matrix: Highly cross-linked 4% agarose
- Binding capacity: >25mg horse liver GST/mL
- pH stability working range: 4 to 13
Specifications
5 mL | |
16 mm | |
<20 mL/min. | |
4 to 13 | |
4°C to 30°C, 20% Ethanol | |
45 to 165 μm | |
GSTrap 4B columns are used for the purification of GST-tagged proteins from bacterial lysates and other glutathione S-tranferases or glutathione-dependent proteins. Mild elution conditions preserve protein antigenicity and function | |
Glutathione |
0.62 in. | |
GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins. | |
Polypropylene | |
4% Highly Cross-linked Agarose | |
5 bar (0.5 MPa, 70 psi) | |
1 x 5 mL | |
Pre-packed Column |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction