Learn More
GE Healthcare High Purity Solution ddNTPs
Use in any application that requires ddNTPs of high purity. GE Healthcare Life Sciences™ High Purity Solution ddNTPs are available either as individual products or in a convenient set.
Manufacturer:  GE Healthcare 27207101
 View more versions of this product
Catalog No. 45-001-344
Add to cart
Description
Description
  • Each lot of ddNTPs is functionally tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase™ DNA polymerase
  • High purity: >98% triphosphate
Dideoxy sequencing, Genotyping reactions
Specifications
Specifications
100mM
Liquid
ddGTP
4μmol (40μL)
Colorless
Dideoxy sequencing, genotyping reactions
≥98%