Learn More
GE Healthcare High Purity Solution ddNTPs Set (ddATP, ddCTP, ddTTP)
Easy-to-use, time-saving solutions for use in any application requiring ddNTPs of high purity
Manufacturer:  GE Healthcare 27204501
Catalog No. 45-001-339
Add to cart
Description
Description
  • Each lot of ddNTPs is functionally tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase™ DNA polymerase
  • High purity: >98% triphosphate
  • In a convenient set containing ddATP, ddCTP, ddGTP, and ddTTP in separate tubes
Dideoxy sequencing, Genotyping reactions
Specifications
Specifications
5mM
Liquid
2',3'-Dideoxyadenosine 5'-Triphosphate, 100mM Solution (ddATP), 2',3'-Dideoxycytidine 5'-Triphosphate, 100mM Solution (ddCTP), 2',3'-Dideoxyguanosine 5'-Triphosphate, 100mM Solution (ddGTP), 2',3'-Dideoxythymidine 5'-Triphosphate, 100mM Solution (ddTTP)
4 x 0.5μm
Colorless
Dideoxy sequencing, genotyping reactions
≥98%