GE Healthcare High Purity Solution ddNTPs Available on GSA/VA Contract for Federal Government customers only.

Use in any application that requires ddNTPs of high purity. GE Healthcare Life Sciences™ High Purity Solution ddNTPs are available either as individual products or in a convenient set.

Manufacturer: GE Healthcare 27206101

 View more versions of this product

Catalog No. 45-001-342

  Check Availability
Add to Cart

    Specifications

    Format Liquid
    For Use With (Application) Dideoxy sequencing, genotyping reactions
    Product Type ddCTP,ddCTP
    Concentration 100mM
    Purity ≥98%
    Quantity 4μmol (40μL)
    Color Colorless

    • Each lot of ddNTPs is functionally tested to meet or exceed criteria for high-quality sequencing with Thermo Sequenase™ DNA polymerase
    • High purity: >98% triphosphate

    Dideoxy sequencing, Genotyping reactions

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit