Learn More
GE Healthcare HiPrep™ HIC FF 16/10 Columns Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  GE Healthcare 28936546
Catalog No. 45-002-078
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
1M NaOH, 3M Ammonium Sulphate, 30% Isopropanol, 70% Ethanol, 10% Ethylene Glycol, 0.5% SDS, 6M Guanidine Hydrochloride, 8M Urea
0.62 in.
HiPrep Phenyl Sepharose 4 FF (low sub) column
Provide fast, preparative separation of Proteins and other biomolecules by hydrophobic interaction chromato graphy (HIC), initial and intermediate purification, removing bulk impurities
100mm
Polypropylene
5 bar
45 to 165μm
4° to 30°C
20mL
16mm
1 x 20mL
3.93 in.
>300cm/hr.
Phenyl
3 to 13
6% Cross-linked Agarose