Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare His GraviTrap™ Kit

For fast and simple purification of histidine-tagged proteins

Manufacturer:  GE Healthcare 28401351

Catalog No. 45-002-016


Add to cart

Includes: Two packs of His GraviTrap and one pack of His buffer kit

Description

Description

  • Binding capacity: approximately 40mg histidine-tagged protein/column
  • Bed volume: 1mL
  • Column hardware material: polypropylene (barrel), polyethylene (frits)

Specifications

Specifications

1mL
His GraviTrap kit
2 packs of His GraviTrap and 1 pack of His Buffer Kit.
His GraviTrap Kit
4 - 30°C, 20% Ethanol
Approx. 40 mg histidine-tagged protein/column
For fast and simple purification of histidine-tagged* proteins
Polypropylene (PP) barrel, Polyethylene (PE) frits
10 Columns
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit