Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva His Mag Sepharose™ excel Chromatography Media

His Mag Sepharose excel magnetic beads are designed for small-scale purification and screening of histidine-tagged proteins secreted in eukaryotic cell culture.

$296.00 - $790.00

Specifications

pH Range 2 to 12
Support Type Highly cross-linked spherical Agarose including Magnetite
Content And Storage 4°C to 30°C
Particle Size 63 μm
For Use With (Application) Designed to simplify small-scale purification and screening of His-tagged Proteins_x000D_ secreted in eukaryotic cell culture, used for functional and structural studies research in academia, institutes and discovery Pharma
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-002-369
View Documents
Cytiva
17371222
10 x 1 mL
Each for $790.00
Add to Cart
 
45-003-016
View Documents
Cytiva
17371221
5 x 1 mL
Each for $455.00
Add to Cart
 
45-003-015
View Documents
Cytiva
17371220
2 x 1 mL
Each for $296.00
Add to Cart
 
Description

Description

Samples that usually cause stripping of metal ions can therefore be added to the medium with retained protein-binding capacity and without pretreatment.

  • His Mag Sepharose excel is a magnetic IMAC medium precharged with nickel ions that are very strongly bound to a chelating ligand.
  • The medium enables an efficient and simplified workflow for capture, purification, and reproducible screening of histidine-tagged proteins.
  • Simplify small-scale purification and expression screening of histidine-tagged proteins secreted in eukaryotic cell culture.
  • Purify histidine-tagged proteins directly from samples that contain metal ion-stripping agents and save time and effort.
  • Increase target protein yield and decrease degradation through reduced and simplified sample handling.
  • Screen histidine-tagged proteins in parallel with high reproducibility and reliable results.
  • Capture histidine-tagged protein from different sample volumes; from low to high milliliter scale.
  • Simplify handling using visible and dense Sepharose-based magnetic beads.
  • Proteins are expressed and secreted into cell culture supernatants from eukaryotic cells such as insect cells or Chinese Hamster Ovary (CHO) cells.
  • The high density of the beads allows rapid capture by magnetic devices while the visibility of the beads ensures reliable collection of the bound histidine-tagged proteins during the purification procedure.
  • The base matrix is based on Sepharose medium with magnetite incorporated.
  • His Mag Sepharose is available as 10% medium slurry in three different pack sizes (2 x 1ml, 5 x 1ml, and 10 x 1ml) and can be used together with different test tubes and magnetic racks.

Specifications

Specifications

2 to 12
4°C to 30°C
Designed to simplify small-scale purification and screening of His-tagged Proteins_x000D_ secreted in eukaryotic cell culture, used for functional and structural studies research in academia, institutes and discovery Pharma
Highly cross-linked spherical Agarose including Magnetite
63 μm
Chromatography Media
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit