Learn More
GE Healthcare His Mag Sepharose™ Ni Magnetic Beads
Designed for efficient, small-scale purification/screening of histidine-tagged proteins from different sources GE Healthcare Lifescience™ His Mag Sepharose™ Ni Magnetic Beads enable high-capacity small-scale purification.
Manufacturer:  GE Healthcare 28979917
 View more versions of this product
Catalog No. 45-002-516
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel. For higher throughput, larger numbers of samples can easily be screened in parallel on a robotic device.
  • High-capacity small-scale purification of histidine-tagged proteins from different sources
  • High purity and yield
  • Easy parallel screening of histidine-tagged proteins with high repeatability
  • Scalability dash (en) Simple capture of histidine-tagged proteins from small or large sample volumes (low microliter to milliliter scale)
Specifications
Specifications
50mg Histidine-tagged protein/mL medium