Learn More
Cytiva His MultiTrap™ 96-well Filter Plates
Prepacked multiwell filter plates for high reproducibility in expression screening and small-scale purification of histidine-tagged proteins
Supplier: Cytiva 28400990
Description
- No filtration needed: load unclarified sample directly, increasing reproducibility and saving time
- Compatible with robotic systems or manual protocols with centrifuge or vacuum
- Easy scale-up
- Long-term pH range 3 to 12
- Protein binding capacity up to 800μg histidine-tagged protein/well with His MultiTrap FF
- Protein binding capacity up to 1mg histidine-tagged protein/well with His MultiTrap HP
Specifications
His MultiTrap FF and His MultiTrap HP are prepacked disposable 96-well plates for reproducible high throughput parallel purification of histidine-tagged recombinant proteins by Immobilized Metal ion Affinity Chromatography (IMAC) | |
3 to 12 | |
4°C to 30°C | |
4 x 96-well Filter Plates |
Polypropylene, Polyethylene | |
6% Cross-linked Agarose | |
45 to 165 μm | |
96-well Filter Plate |