Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare HisTrap™ FF Columns: 7 x 25mm Promotion

For a limited time only, receive a 20% discount on your next HiTrap order

Manufacturer:  GE Healthcare 17525501PM/DEL

Catalog No. 45-000-326PM

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Prepacked with precharged Ni Sepharose™ 6 Fast Flow media
  • Column material: polypropylene
  • Bed dimensions: 7 x 25mm
  • Protein binding capacity of ~40mg/mL media with negligible nickel ion leakage
  • Compatible with wide range of reducing agents, detergents, denaturants, and other additives
  • Simple operation with syringe, perstaltic pump, or chromatographic system
  • Columns may be connected in series to increase capacity
Medium
  • Matrix: 6% cross-linked agarose
  • Average particle size: 90µm
  • pH stability working range: 3 to 12

  • Cannot be combined with any other discounts
  • Offer expires June 25, 2015

Specifications

Specifications

5mL
0.2 in.
<4mL/min.
5 × 5mL
HisTrap Column
5 × 5mL
6% Cross-linked Agarose
≈40mg His-tagged protein/column
7mm
Polypropylene
3 to 12
5 bar
4° to 30°C in 20% ethanol
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit