Learn More
GE Healthcare HisTrap™ HP Prepacked Columns Promotion
For a limited time only, receive a 20% discount on your next HiTrap order
Manufacturer:  GE Healthcare 29334694
 View more versions of this product
Catalog No. 45-000-324PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Specifications
5mL
0.6 in.
5mL/min.
1 x 5mL
HisTrap HP Column
1 x 5mL
6% Highly Cross-linked Agarose
≈40mg His-tagged Protein/mL
16mm
Polypropylene
3 to 12
3 bar
4° to 30°C