Learn More
GE Healthcare HiTrap™ Chelating HP Columns Available on GSA/VA Contract for Federal Government customers only.
Optimize histidine-tagged protein purifications when nickel ions are not the best choice. GE Healthcare Life Sciences™ HiTrap™ Chelating HP Columns is prepacked with Chelating Sepharose™ High Performance.
Manufacturer:  GE Healthcare 17040905
 View more versions of this product
Catalog No. 45-002-910
  Check Availability
Add to cart
Specifications
Description
  • Uses different kinds of metal ions
  • Binding capacity ~12mg histidine-tagged protein/mL medium
  • Metal ion capacity ~23µmol Cu2+/mL medium
  • Prepacked with Chelating Sepharose High Performance media
  • Iminoacetic acid coupled to a 6% agarose matrix by ether linkage
Description & Specifications
Specifications
6M Guanidine Hydrochloride, 8M Urea
HiTrap chelating HP column
IMAC and subsequent purification of Polyhistidine tagged Proteins
25mm
Iminiodiacetic acid chelating group
3 bar
34μm
4° to 30°C
5mL
100 x 5mL
0.98 in.
<20mL/min.
Iminodiacetic Acid
3 to 13
6% Highly Cross-linked Agarose