GE Healthcare™ GE Healthcare HiTrap™ Desalting and HisTrap™ Pre-packed Columns: Promo Offer

Special offer is available 1/1/2016 through 3/31/2016

Manufacturer: GE Healthcare™ 29180134

 View all options for this product

Catalog No. 45-000-511PM


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Description Packaging Quantity: 1 x 5mL
    Quantity Each
    Product Type HiTrap Desalting Columns
    Particle Size >20um (Dry), 15 to 88um (wet)
    Diameter (English) Inner Column 0.62 in.
    For Use With (Application) For rapid convenient desalting and buffer exchange
    Diameter (Metric) Inner Column 1.6cm
    Storage Requirements 4 to 30°C
    Volume (Metric) Bed 5mL

    •  Sephadex G-25 superfine media
    • Non-disposable

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit