Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare™ GE Healthcare HiTrap™ Desalting and HisTrap™ Pre-packed Columns: Promo Offer

Special offer is available 1/1/2016 through 3/31/2016

Manufacturer:  GE Healthcare™ 29180135

 View more versions of this product

Catalog No. 45-000-252PM

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

  • Material: Sephadex G-25 superfine media
  • Non-disposable
Description & Specifications

Specifications

Packaging Quantity: 5 x 5mL
HiTrap Desalting Columns
0.62 in.
1.6cm
5mL
5 Pack
>20um (Dry), 15 to 88um (wet)
For rapid convenient desalting and buffer exchange
4 to 30°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit