GE Healthcare HiTrap™ Desalting Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.

For rapid convenient desalting and buffer exchange. HiTrap™ Desalting Prepacked Columns are prepacked, ready to use column for group separation between high and low molecular weight substances.

Manufacturer: GE Healthcare 11000329

 View more versions of this product

Catalog No. 45-002-908

  Check Availability
Add to Cart

    Specifications

    Disposable No
    Particle Size >20μm (Dry), 15 to 88μm (wet)
    Diameter (English) Inner Column 0.62 in.
    For Use With (Application) For rapid convenient desalting and buffer exchange
    Diameter (Metric) Inner Column 1.6cm
    Storage Requirements 4° to 30°C
    Volume (Metric) Bed 5mL
    Quantity 100 Pack

    • Prepacked with Sephadex™ G-25 Superfine media
    • Column material: polypropylene
    • Bed volume: 5mL
    • Bed dimensions: 16 x 25mm
    • Sample volume: 1.5mL
    • Flow velocity: <15mL/min.
    • Maximum pressure: 3 bar

    Media
    • Matrix: cross-linked dextran
    • Partical size: >20µm
    • pH stability working range: 2 to 13

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit