GE Healthcare HiTrap™ MabSelect™ Prepacked Columns Promotion

For a limited time only, receive a 20% discount on your next HiTrap order

Manufacturer: GE Healthcare 29133597

 View all options for this product

Catalog No. 45-000-010PM


This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Quantity 5 Pack
    Diameter (Metric) Inner Column 1.6cm
    Diameter (English) Inner Column 0.62 in.
    Binding Capacity ≈35mg Human lgG/mL at 2.4 min. residence time
    Flow Rate <20mL/min.
    For Use With (Application) For fast purification of monoclonal antibodies from large sample volumes, affinity purification
    Storage Buffer 20% Ethanol
    Storage Requirements 4° to 30°C
    Sample Volume 5mL
    Volume (Metric) Bed 5mL

    Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs. MabSelect media are suited for the high flowrates and pressures required when scaling up.

    • Oriented coupling of recombinant Protein A ligand to the agarose matrix via an engineered C-terminal cysteine increases IgG binding capacity over other protein A media
    • High capacity, low ligand leakage, and high-flow base matrix allow purification of Mabs at process scale
    • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
    • Designed for use with a syringe, a peristaltic pump, or a chromatography system


    • Allows at least five times higher flowrates than conventional cross-linked agarose of similar porosity
    • Recombinant Protein A ligand promotes specificity of binding to Fc region of IgG; permits one-step purification
    MabSelect Xtra™
    • Same recombinant protein A ligand as MabSelect, but with a smaller particle size and greater porosity, for increased dynamic binding capacity
    • Allows binding from samples with high expression levels of the monoclonal antibody
    • Lowers overall production cost due to the possibility of processing concentrated feedstocks in fewer batches
    MabSelect SuRe™ (Superior Resistance)
    • Akali-tolerant protein A-derived ligand allows the use of up to 0.5M NaOH for cleaning-in-place (CIP)
    • Permits significant cost savings and extension of cycles in regular production

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit