GE Healthcare HiTrap™ Protein G HP Prepacked Columns Promotion

For a limited time only, receive a 10% discount on your next HiTrap order

Manufacturer: GE Healthcare 29-1335-88

 View all options for this product

Catalog No. 45-000-056PM

  Check Availability
Add to Cart


    Quantity 5 Pack
    Binding Capacity 125mg Human IgG/column
    Diameter (Metric) Inner Column 1.6cm
    Diameter (English) Inner Column 0.62 in.
    For Use With (Application) Fast, easy separations of IgG from different species and subclasses.
    Flow Rate <20mL/min.
    Storage Buffer 20% Ethanol
    Storage Requirements 4° to 30°C
    Volume (Metric) Bed Bed Dimensions: 16 x 25mm; 5 x 5mL


    • Prepacked with Protein G Sepharose High Performance
    • Binding capacity: 25mg human IgG/column
    • Maximum pressure: 3 bar
    • Column material: polypropylene
    • For use with syringe, peristalic pump, or chromatography system
    • 1 and 5mL columns
    • Ligand: Recombinant protein G lacking albumin-binding region
    • Matrix: 6% cross-linked agarose
    • Average particle size: 34µm
    • pH stability working range: 3 to 9



    • Cannot be combined with any other discounts
    • Offer expires June 30, 2016


    1mL columns

    • Bed dimensions: 7 x 25mm
    • Flow velocity: <4mL/min.
    5mL columns
    • Bed dimensions: 16 x 25mm
    • Flow velocity: <20mL/min.


  • Description & Specifications
  • Promotions

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit