GE Healthcare HiTrap™ rProtein A FF Columns

For a limited time only, receive a 10% discount on your next HiTrap order

Manufacturer: GE Healthcare 29-1335-76

 View more versions of this product

Catalog No. 45-000-262PM

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Quantity 5 x 1mL
    Binding Capacity ≈50mg Human IgG/mL
    Flow Rate <4mL/min.
    For Use With (Application) For preparative purification of monoclonal and polyclonal antibodies from cell culture supernatants, Serum and ascites
    For Use With (Equipment) For syringe, peristaltic pump or a chromatography system
    Storage Requirements 4° to 30°C

     

    • Packed with rProtein A Sepharose™ Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources
    • Exhibits similar Fc region specificity to that of native protein A but with enhanced binding capacity and fewer regulatory concerns due to total absence of mammalian culture in ligand production and purification
    • Average particle size: 90µm
    • Designed for use with a syringe, a peristaltic pump, or a chromatography system
    • Available as 1mL and 5mL columns
    Media
    • Ligand: Recombinant protein A (E.coli)
    • Matrix: Highly cross-linked 4% agarose
    • Binding capacity: ∽50mg human IgG/mL
    • pH stability working range: 3 to 10

     

     

    • Cannot be combined with any other discounts
    • Offer expires June 30, 2016

     

    1mL columns

    • Bed dimensions: 7 x 25mm
    • Flow velocity: <4mL/min.
    5mL columns
    • Bed dimensions: 16 x 25mm
    • Flow velocity: <20mL/min.

     

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit