GE Healthcare HiTrap™ rProtein A FF Columns

For a limited time only, receive a 10% discount on your next HiTrap order

Manufacturer: GE Healthcare 29133582

 View more versions of this product

Catalog No. 45-000-265PM

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Quantity 5 x 5mL
    Binding Capacity ≈50mg Human IgG/mL
    Flow Rate <20mL/min.
    For Use With (Application) For preparative purification of monoclonal and polyclonal antibodies from cell culture supernatants, Serum and ascites
    For Use With (Equipment) For syringe, peristaltic pump or a chromatography system
    Storage Requirements 4° to 30°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit