Learn More
Hoefer™ PR 648 Slot Blot Blotting Manifold Available on GSA/VA Contract for Federal Government customers only.
Provides a simple, convenient method for screening recombinant clones, DNA, RNA and proteins
Manufacturer:  Hoefer™ PR648
 View more versions of this product
Catalog No. 03-500-440
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Slot Blot Blotting Manifold
3.14 in.
12cm
15.5cm
Top block, membrane support block with O-ring, bottom block with connector, QuickFit™ connector with O-ring, 6 clamping screws, membrane cutting template
48.0
45°C
4.72 in.
6.1 in.
8cm
3.14 x 6.1 x 4.72 in. (8 x 15.5 x 12cm)
Acrylic
113°F
1mL
Accessories