Learn More
GE Healthcare HR (High Resolution) Columns
HR (High Resolution) columns are for large-scale applications with high-performance media.
Manufacturer: GE Healthcare 19740301
Description
- Compatible with any high performance system including ÄKTA design systems, HR columns provide precision sample application and elution, and excellent physical and chemical stability.
- HR 16 columns are available in three different lengths: 5, 10, and 50 cm
- Glass construction, with no metal parts, allows visual observation at all times.
- Wide range of accessories, adapters, and replacement parts available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
14.3-22.3mL (with one adapter), 6.2-22.3mL (with 2 adapters) | |
HR (High Resolution) Chromatography Columns | |
Borosilicate glass |
16mm | |
100mm | |
430psi (30 bar) |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.