Learn More
GE Healthcare HR (High Resolution) Columns Available on GSA/VA Contract for Federal Government customers only.
HR (High Resolution) columns are for large-scale applications with high-performance media.
Manufacturer:  GE Healthcare 18100098
 View more versions of this product
Catalog No. 45-200-33
Add to cart
Description
Description
  • Compatible with any high performance system including ÄKTA design systems, HR columns provide precision sample application and elution, and excellent physical and chemical stability.
  • HR 16 columns are available in three different lengths: 5, 10, and 50 cm
  • Glass construction, with no metal parts, allows visual observation at all times.
  • Wide range of accessories, adapters, and replacement parts available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
5.4-13.7mL (with one adapter), 0-13.9mL (with 2 adapters)
HR (High Resolution) Chromatography Columns
Borosilicate glass
16mm
50mm
430psi (30 bar)