Learn More
GE Healthcare illustra™ bacteria genomicPrep Mini Spin Kit
Designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria
Manufacturer:  GE Healthcare 28904258
 View more versions of this product
Catalog No. 45-001-493
Add to cart
Includes: illustra bacteria genomicPrep Mini spin columns prepacked with novel silica membrane; Lyophilized proteinase K powder; Lysis solutions, wash and elution buffers; 2mL Microcentrifuge collection tubes; a full booklet with a detachable, quick reference protocol card
Description
Description
The illustra Bacteria genomicPrep Mini Spin Kit can be used with a wide range of bacterial strains such as DH5a, TOP10, JM109, Bacillus subtilis etc. The kit produces highly pure and intact genomic DNA with OD260/280 values of 1.75-1.85. Use different elution volumes to isolate genomic DNA at concentrations suitable for most downstream applications including restriction enzyme digestion, RT-PCR, and long PCR.
The method uses a lysis solution in combination with proteinase K to gently release genomic DNA into solution from bacteria cells. The DNA is deproteinated in the extraction solution and the genomic DNA is then bound onto a silica column in the presence of chaotropic solution. Contaminants are removed during wash steps and the purifed DNA is eluted with prewarmed TE solution.
  • Total DNA extraction time under one hour with reduced lysis time
  • Can be used to purify 4 to 8µg of genomic DNA from 2 x 109 bacteria cells with input amounts ranging from 1 x 109 to 4 x 109 bacteria cells
  • Spin columns prepacked with a silica membrane—provide excellent reproducibility and consistency
  • Validated in several downstream applications including RT PCR, long PCR, and restriction digest
  • Color-coded caps for ease of use
Specifications
Specifications
illustra bacteria genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from Gram-negative (G-) and Gram-positive (G+) bacteria.
2 x 30mg Proteinase K, 15mL Lysis Buffer type 2, 2 x 1.5mL Lysis Buffer type 3, 2 x 165mL Lysis Buffer type 4, 30mL Wash Buffer type 6, 60mL Elution Buffer type 5, 5 x 50 Illustra™ bacteria mini columns, 5 x 50 collection tubes
50 Preps
2 x 109 cells [A600 = 2.0]
For the rapid extraction and purification of high-quality genomic DNA from gram-positive and gram-negative bacteria Purified gDNA is suitable for molecular biology applications including cloning, restriction enzyme digestion, PCR and genotyping
illustra bacteria genomicPrep Mini Spin Kit
40 min [Gram-negaitive Bacteria], 55 min [Gram-positive Bacteria]
1.7