Learn More
GE Healthcare illustra™ ExoProStar™ S PCR and Sequence Reaction Clean-up Kit Available on GSA/VA Contract for Federal Government customers only.
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
Manufacturer:  GE Healthcare US79010
 View more versions of this product
Catalog No. 45-002-351
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product
PCR and Sequence Reaction Cleanup kit
100 Reactions