Learn More
GE Healthcare illustra™ ExoProStar™ S PCR and Sequence Reaction Clean-up Kit
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
Manufacturer:  GE Healthcare US79500
 View more versions of this product
Catalog No. 45-002-354
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product
PCR and Sequence Reaction Cleanup kit
5000 Reactions