GE Healthcare illustra™ mRNA Purification Kit
For rapid affinity purification of polyadenylated mRNA from eukaryotic total RNA using prepacked oligo(dT)-cellulose spin columns
Manufacturer: GE Healthcare 27925801
 View more versions of this product
Catalog No. 45-500-120
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Application) cDNA synthesis, PCR, northern blot hybridization and in-vitro translation, for rapid affinity purification of mRNA from eukaryotic total RNA using prepacked Oligo(dT)-cellulose spin columns
    Format Spin column, filter column
    Includes Oligo(dT)-cellulose columns 4, high salt Buffer 2 vials, sample Buffer 2 vials, low salt Buffer 2 vials, Elution Buffer 2 vials, Glycogen solution 1 vial, protocol 1
    Quantity 2 Preps
  • Description & Specifications
Related Products
Product Content Feedback