Learn More
GE Healthcare illustra™ Ready-To-Go™ GenomiPhi HY DNA Amplification Kit Available on GSA/VA Contract for Federal Government customers only.
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60++g yield from nanogram amounts of DNA sample.
Manufacturer:  GE Healthcare 25660397
 View more versions of this product
Catalog No. 45-003-106
Add to cart
Description
Description
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from GE Healthcare.
  • Representative amplification of the whole genome.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
  • Quick and simple automation-friendly protocol; no thermal cycler required.
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint.
  • Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample.
5 x 2.6mL 2X Denaturation Buffer, 5 x 96-well strips Ready-To-Go™ GenomiPhi HY cakes, 1 x vial control DNA, 12 strips of 8 domed caps
480 Reactions
4 h
Genomic DNA Amplification
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit 5x96 Reactions
Store the kit at ambient temperature. Once opened, completely reseal the pouch containing the Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended. Store recons
> 10 kb