Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare illustra™ Solution dNTPs

Use dNTPs incorporating the most recent advances in manufacturing technology. GE Healthcare illustra™ Solution dNTPs offer the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

Manufacturer:  GE Healthcare 28406523

 View more versions of this product

Catalog No. 45-001-448

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

 

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing

 

Description & Specifications

Specifications

100mM
Liquid
7
>99%
Colorless
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
dGTP
500μm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit