Learn More
GE Healthcare illustra™ Solution dNTPs Available on GSA/VA Contract for Federal Government customers only.
Use dNTPs incorporating the most recent advances in manufacturing technology. GE Healthcare illustra™ Solution dNTPs offer the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.
Manufacturer:  GE Healthcare 28406503
 View more versions of this product
Catalog No. 45-500-122
Add to cart
Description
Description
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Custom formulation, testing, packaging, and labeling are available
Specifications
Specifications
100mM
Liquid
7
500μm
Colorless
Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
>99%