GE Healthcare illustra™ Solution dNTPs

Use dNTPs incorporating the most recent advances in manufacturing technology. GE Healthcare Life Sciences™ illustra™ Solution dNTPs offer the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

Manufacturer: GE Healthcare 28406531

 View more versions of this product

Catalog No. 45-001-449

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Format Liquid
    For Use With (Application) Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling
    pH 7
    Product Type dTTP,dTTP
    Concentration 100mM
    Purity >99%
    Quantity 25μm
    Color Colorless

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit