Learn More
GE Healthcare Immunoprecipitation Starter Pack
Two different media, nProtein A Sepharose™ Fast Flow and Protein G Sepharose Fast Flow, permit affinity capture of antibodies from a wide range of mammalian species allowing effective and rapid removal of immune complexes via immunoprecipitation
Manufacturer:  GE Healthcare 17600235
Catalog No. 45-000-369
Add to cart
Includes: Each unit includes 2mL nProtein A Sepharose 4 Fast Flow and 2mL Protein G Sepharose 4 Fast Flow with instructions for analytical separations of target antigens from crude cell lysates. Sold in packs of two units.
Description
Description
  • High specificity and high binding capacity with minimal nonspecific adsorption and low ligand leakage
  • Binding capacity nProtein A, >30mg human IgG/mL medium; Protein G, >20mg human IgG/mL medium Step-by-step instructions guide you through the procedure
  • Inclusion of both protein A and protein G expands the species spectrum improving flexibility, yield and selectivity
  • Each unit is sufficient for approximately 80 immunoprecipitations
  • Long term pH stability: 3-9
Specifications
Specifications
For analytical separations of target antigens
1 x 2mL nProtein A Sepharose 4 fast flow, 1 x 2mL ProteinG Sepharose 4 fast flow, instructions
2 x 2mL
4° to 30°C
Liquid
Immunoprecipitation starter pack
Cell Lysate
80 Preps
Safety and Handling
Safety and Handling
ShelfLife : 3 Year