GE Healthcare IPG Buffers Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: GE Healthcare 17600202

Catalog No. 45-000-363

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Format Liquid
    pH 3 to 5.6 nonlinear
    Product Name IPG Buffer
    Quantity 1mL
    Storage Requirements 2° to 8°C
    For Use With (Application) Used for isoelectric focusing staining in 2-D Electrophoresis, eliminates background staining, improve Protein solubility, ensures optimal conditions for IEF separation
    For Use With (Equipment) Immobiline DryStrip Gels
    Color Colorless to Light Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit