Learn More
GE Healthcare Low Fluorescence Glass Plate Kit
Manufacturer:  GE Healthcare 80647558
Catalog No. 45-005-915
Add to cart
Includes:
  • One borosilicate glass plate (27.6 x 21.7cm x 3mm)
  • One borosilicate glass plate (27.6 x 21.1cm x 3mm)
  • Two vinyl spacer (10mm x 1mm)
Specifications
Specifications
1 Borosilicate Glass plate (27.6 x 21.7cm x 3mm), 1 Borosilicate Glass plate (27.6 x 21.1cm x 3mm), 2 Vinyl spacer (10 x 1mm)
Complete packsize
Low Fluorescence Glass Plate Kit