Learn More
GE Healthcare Mammalian Protein Extraction Buffer
After preparation may be used for most enzyme assays
Manufacturer:  GE Healthcare 28941279
Catalog No. 45-002-169
Add to cart
Includes: <20mM tris hydroxymethyl aminomethane, <20mM sodium chloride, <5% nonylphenyl polyethylene glycol, <5% octylphenolpoly(ethyleneglycolether), and <5% polyethylene glycol sorbitan monolaurate
Description
Description
This buffer is based on organic buffering agents, mild nondenaturing buffer, compositions and can be used for both cell suspensions as well as adherent cells. Mammalian Protein Extraction Buffer is compatible with most applications, including enzyme assays, various chromatography procedures, and electrophoresis. After preparation, the sample may be used for most enzyme assays including reporter gene assays, kinases, and immunoassays.
Specifications
Specifications
<20mM Tris Hydroxymethyl Aminomethane, <20mM Sodium Chloride, <5% Nonylphenyl Polyethylene Glycol, <5% Octylphenolpoly (Ethyleneglycolether) and <5% Polyethylene Gly
Mammalian Protein Extraction Buffer
Instructions supplied with the buffer
500 mL
Enzyme assays, various chromato graphy procedures and electrophoresis, efficient and gentle extraction of biologically active, to tal soluble Proteins from mammalian cultured cells
Storage temperature: 2–8°C