Learn More
GE Healthcare MiniBeads™ Columns
MiniBeads™ prepacked columns are designed for high performance purification and characterization of biomolecules using ion exchange chromatography
Manufacturer:  GE Healthcare 17517801
 View more versions of this product
Catalog No. 45-000-308
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid
0.18 in.
Mini S ion exchange column
High performance purification and characterization of biomolecules by ion exchange chromato graph
50mm
Valco fittings
O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-SO3
3 to 11
Monodisperse Polystyrene/Divinyl Benzene particles
0.8mL
4.6mm
Each
1.96 in.
0.5 to 2mL/min.
Polyethere Ketone
180psi
3μm
4° to 30°C