Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Mono Q PC, Columns Prepacked with MonoBeads™
Use Mono Q or S precision columns for fast, sensitive, and high-resolution ion exchange separations of biomolecules at micropreparative scale. Cytiva Life Sciences™ Mono Q or S PC, Columns Prepacked with MonoBeads™ achieve maximum resolution from the efficiency of small, perfectly spherical monodispersed particles.
Supplier: Cytiva 17516701
Description
- Maximum resolution achieved from efficiency of small spherical particles, optimally packed
- Particles are coupled to Q or S ion exchangers with excellent sensitivities
- Compatible with high-performance LC systems
- Stable in all common buffers except high-concentration oxidizing and reactive agents, DMSO, DMF and formic acid
- Tricorn columns are optimized for the polishing step when high purity is required
- PC 1.6/5 columns are excellent for micropurification of subnanogram to microgram amounts of sample on high-performance LC systems
- Mono Q: ionic capacity 0.27 to 0.37mmol Cl-/mL
- Mono S: ionic capacity 0.12 to 0.15mmol Cl-/mL
Specifications
All common Buffers except high-concentration Oxidizing and reactive agents, DMSO, DMF and Formic Acid | |
8mL | |
10mm | |
3.93 in. | |
2 to 6mL/min. | |
O-CH2-CHOH-CH2-O-CH2-CHOH-CH2-N+ (CH3) 3 | |
2 to 12 | |
Monodisperse porous Polystyrene/Divinyl Benzene bead | |
Mono Q Ion Exchange Column |
Mono Q ion exchange column | |
0.39 in. | |
Each | |
100mm | |
Borosilicate Glass | |
40 bar | |
10μm | |
4° to 30°C | |
Small-scale polishing in purification of Proteins, Peptides and other biomolecules when high purity is required |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction