Learn More
GE Healthcare Mono P Columns
Greatly concentrates samples and resolves pI differences of 0.02 pH units
Manufacturer:  GE Healthcare 17517101
 View more versions of this product
Catalog No. 45-000-306
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
4mL
Borosilicate Glass
Each
Monodisperse polystyrene/ divinyl benzene
Rapid scouting of elution conditions (pH gradients) or for separations not requiring highest resolution
Mono P ion exchange column
4° to 30°C