Learn More
GE Healthcare NHS HP SpinTrap™ Buffer Kit
Pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation according to the NHS SpinTrap column protocol
Manufacturer:  GE Healthcare 28913569
Catalog No. 45-001-524
Add to cart
Includes: 10x Binding/Washing Buffer (TBS)(0.5 M Tris, 1.5 M NaCl, pH 7.5); 10x Elution Buffer (1 M glycine-HCl, pH 2.9); 5x Coupling Buffer (0.75 M triethanolamine, 2.5 M NaCl, pH 8.3); 5x Blocking Buffer A (2.5 M ethanolamine, 2.5 M NaCl, pH 8.3); 5x Blocking Buffer B (0.5 M sodium acetate, 2.5 M NaCl, pH 4.0)
Description
Description
  • High quality buffers
  • All buffer components defined
  • Convenient format- stock solutions can be diluted directly into the provided bottles for storage
  • Eliminates time-consuming buffer preparation, which allows fast, reproducible protein enrichment
  • Pre made buffers for use with NHS SpinTrap HP columns
Specifications
Specifications
Buffer kit
Immunoaffinity enrichment using the NHS HP SpinTrap
4° to 30°C
24 Reactions
3 to 12