GE Healthcare NovaSpec™ Plus Visible Spectrophotometer Available on GSA/VA Contract for Federal Government customers only.

Incorporates diode array technology for rapid wavelength scanning in a highly reliable and low-maintenance instrument suitable for general biotech laboratory use

Manufacturer: GE Healthcare 80211751

 View more versions of this product

Catalog No. 45-001-652

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Visible
    Width (Metric) 31cm
    Detector Type Diode array
    Electrical Requirements 90/265V 50/60Hz
    Includes Grafico PC utility software, serial cable, dust cover
    Wavelength Range 330 to 800nm
    Certifications/Compliance CE 89/336/EEC (EMC directive); CE 73/23/EEC (LV directive); EN-61010-1 (IEC1010-1)
    Weight (English) 3 lb.
    Depth (English) 9.05 in.
    For Use With (Application) Basic enzyme kinetics, bradford, BCA, biuret and lowry protein quantitation
    Height (Metric) 13cm
    Hertz 50/60Hz
    Lamp Tungsten Halogen
    Spectral Bandwidth 7nm
    Voltage 90/265V
    Wavelength Accuracy ±2nm
    Weight (Metric) 1.75kg
    Width (English) 12.2 in.
    Depth (Metric) 23cm
    Height (English) 5.11 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit