Learn More
GE Healthcare NovaSpec™ Plus Visible Spectrophotometer
Incorporates diode array technology for rapid wavelength scanning in a highly reliable and low-maintenance instrument suitable for general biotech laboratory use
Manufacturer:  GE Healthcare 80211750
 View more versions of this product
Catalog No. 45-001-651
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Visible
9.05 in.
Diode array
Basic enzyme kinetics, bradford, BCA, biuret and lowry protein quantitation
13cm
Grafico PC utility software
10mm
90/265V
330 to 800nm
1.75kg
31cm
CE 89/336/EEC (EMC directive); CE 73/23/EEC (LV directive); EN-61010-1 (IEC1010-1)
23cm
90/265V 50/60Hz
5.11 in.
50/60Hz
Tungsten Halogen
7nm
±2nm
3 lb.
12.2 in.