GE Healthcare nProtein A Sepharose™ 4 Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.

Used for purification of monoclonal and polyclonal antibodies.   GE Healthcare Lifescience™ nProtein A Sepharose™ 4 Fast Flow Affinity Media contains native protein A coupled to Sepharose 4 Fast Flow.

Manufacturer: GE Healthcare 17528002

 View more versions of this product

Catalog No. 45-002-982

  Check Availability
Add to Cart

    Specifications

    Binding Capacity >35mg Human IgG/mL
    Flow Rate 150 to 250cm/Hour
    For Use With (Application) For the purifiCatIon of monoclonal and polyclonal antibodies at both laboratory and process scale
    Height (English) Bed 9.84 in.
    Height (Metric) Bed 25cm
    Ligand Type Native Protein A (S. aureus)
    Matrix 4% Cross-Linked Agarose
    Max. Pressure 100kPa
    Quantity 200mL
    Stability No significant change in chromatographic performance after 1 week storage using 8M Urea, 6M Gua-HCI, 2% Benzyl Alcohol or 20% Ethanol
    Storage Buffer 20% Ethanol
    Storage Requirements 2 to 8°C
    Particle Size 90μm
    Product Type nProtein A Sepharose 4 fast flow media
    pH Range 2 to 10

    • Low leakage of protein A with nearly twice total IgG binding capacity of Protein A Sepharose CL-4B
    • Used in large-scale FDA-approved processes
    • Manufactured without using animal-derived components
    • Rigidity and high degree of substitution of Sepharose 4 Fast Flow matrix facilitates speedy process of large volumes dilute cell culture fluid
    • High chemical, thermal and mechanical stability
    • Withstands high concentrations of hydrogen bond disrupting agents

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit