Learn More
GE Healthcare Nuclease Mix
Specifically developed for removal of nucleic acids in sample preparation for IEF/2-D electrophoresis applications
Manufacturer:  GE Healthcare 80650142
Catalog No. 45-001-741
Add to cart
Description
Description
Removal of nucleic acids is often required to avoid contamination and subsequent artifacts on 2-D gels. The Nuclease Mix offers an effective mix of DNase and RNase enzymes, as well as the necessary cofactors for optimal nuclease activity.
  • Compatible with GE Healthcare Protease Inhibitor Mix
  • Ready to use: add mix to sample and incubate
  • Does not contain EDTA, a known inhibitor of nucleases
Proteomics
Specifications
Specifications
Nuclease Mix
0.5mL
Removal of Nucleic Acids in sample preparation for IEF/2-D electrophoresis applications in proteomic studies
-20°C