Learn More
GE Healthcare illustra Nucleon™ HT Genomic DNA Extraction Kit
Phenol-free extraction of genomic DNA from paraffin-embedded sections and hard tissue requiring proteinase K digestion. Consistent extraction of high-quality, amplifiable DNA. Protocols developed for those tissues which don't homogenize easily in lysis buffer and require digestion with proteinase K.
Manufacturer:  GE Healthcare RPN8509
Catalog No. 45-000-957
Add to cart
Includes: Reagent B, 18mL; 5M Sodium perchlorate, 6mL; Nucleon resin, 8mL; Proteinase K, 10mg
Description
Description
  • Extract DNA from FFPE samples, mouse tail, Xiphisternum etc.
  • User-friendly protocol enables efficient processing of large numbers of samples
  • Patented Nucleon resin delivers highly purity by binding macromolecules other than DNA
  • DNA is not bound to any solid surface throughout the purification avoiding exposure to shearing forces promoting recovery of high yields of highly intact DNA
  • Each kit is designed to recover high-quality DNA from 50 preparations of up to 25mg of hard tissue or 50 paraffin sections
Specifications
Specifications
illustra Nucleon HT enables DNA extraction from hard tissue and paraffin sections.
18mL Reagent B, 6mL 5M Sodium Perchlorate, 8mL Nucleon™ Resin, 10mg Proteinase K
1 Kit
Extract genomic DNA from Paraffin embedded sections and hard tissue requiring Proteinase K digestion
illustra Nucleon HT
Paraffin-embedded tissue or difficult samples