GE Healthcare Octyl Sepharose™ 4 Fast Flow Chromatography Media

Aliphatic hydrophobic interaction chromatography (HIC) medium for capture and intermediate purification of larger proteins

Manufacturer: GE Healthcare 17094610

 View more versions of this product

Catalog No. 45-002-936

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) For capture/intermediate purification of large proteins
    Ligand Type Octyl
    Material Resin on cross-linked 4% agarose
    Matrix 4% Cross-Linked Agarose
    Max. Pressure 100kPa
    Quantity 25mL
    Stability Stable in commonly used aqueous Buffers: 1M NaOH, 30% Isopropanol, 70% Ethanol, 6M Guanidine-Hydrochloride, 30% Acetonitrile, 1mM HCl
    Storage Buffer 20% Ethanol
    Storage Requirements 4 to 30°C
    Particle Size 45 to 165μm
    Product Type Octyl Sepharose 4 Fast Flow Media
    pH Range 2 to 14

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit